missing translation for 'onlineSavingsMsg'
Learn More

GPR35 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55248-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886745

  • 1656.73 SEK / 20µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to GPR35.

GPCR GPR116 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.
Specifications

Specifications

GPR35
Polyclonal
Unconjugated
GPR35
G protein coupled receptor 35, GPR35, KYNA receptor, Kynurenic acid receptor
Rabbit
Antigen Affinity Chromatography
RUO
2859
-20°C
Liquid
Western Blot, Flow Cytometry
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HC97
GPR35
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers