missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR35 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GPR35 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886745
|
Proteintech
55248-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16876745
|
Proteintech
55248-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to GPR35.
GPCR GPR116 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.Specifications
GPR35 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
Q9HC97 | |
2859 | |
Peptide | |
Primary | |
-20°C | |
GPR35 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G protein coupled receptor 35, GPR35, KYNA receptor, Kynurenic acid receptor | |
GPR35 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |