missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRIN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20212-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to GPRIN3.
May be involved in neurite outgrowth.Specifications
GPRIN3 | |
Polyclonal | |
Unconjugated | |
GPRIN3 | |
GPRIN family member 3, GPRIN3, GRIN3, KIAA2027 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243385, 285513, 502784 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZVF9, Q8BWS5 | |
GPRIN3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GPRIN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction