missing translation for 'onlineSavingsMsg'
Learn More

Munktell Filtrak™ Grade 1F Qualitative High Purity Lab Filter Papers

For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate.

Brand:  Munktell Filtrak™ 110024

 View more versions of this product

Product Code. 11363054

  • 610.00 SEK / Pack of 5
Only null left
Estimated Shipment: 28-07-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of pure cotton linters
  • For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate
  • Basis Weight: 80g/m2
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Disc
5 to 6 ÎĽm
1F
Cotton Linters
29s/10 mL
24 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Munktell Filtrak™ Grade 1F Qualitative High Purity Lab Filter Papers > Diameter: 240 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.