missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Munktell Filtrak™ Grade 5 Qualitative High Purity Lab Filter Papers
For coarse and voluminous precipitates such as iron-aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis.
Brand: Munktell Filtrak™ 110083
Additional Details : Weight : 0.07400kg
Description
- Made of pure cotton linters
- For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis
- Basis Weight: 130g/m2
Specifications
Disc | |
<20 μm | |
5 |
Cotton Linters | |
9s/10 mL | |
110 mm |