missing translation for 'onlineSavingsMsg'
Learn More

Munktell Filtrak™ Grade 5 Qualitative High Purity Lab Filter Papers

For coarse and voluminous precipitates such as iron-aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis.

Brand:  Munktell Filtrak™ 110085

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 11373094

  • 381.00 SEK / Pack of 50
In Stock
to see stock.
Add to basket

Description

Description

  • Made of pure cotton linters
  • For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis
  • Basis Weight: 130g/m2
Specifications

Specifications

Disc
<20 μm
5
Cotton Linters
9s/10 mL
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers