Learn More
Munktell Filtrak™ Grade 5 Qualitative High Purity Lab Filter Papers
For coarse and voluminous precipitates such as iron-aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis.
Brand: Munktell Filtrak™ 110086
This item is not returnable.
View return policy
Description
- Made of pure cotton linters
- For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis
- Basis Weight: 130g/m2
Specifications
Circle | |
<20μm | |
5 |
Cotton Linters | |
9s/10 mL | |
185 mm |