Learn More
Granzyme K Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Granzyme K |
---|---|
Concentration | 500 ÎĽg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885895
|
Proteintech
27894-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16875895
|
Proteintech
27894-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Granzyme K antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Granzyme K. This antibody recognizes human antigen. The Granzyme K antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Granzyme K | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
500 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
3003 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.