missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granzyme M Monoclonal Antibody (4B2G4), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 12-9774-42
This item is not returnable.
View return policy
Description
Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.
Specifications
Granzyme M | |
Monoclonal | |
5 ÎĽL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P51124 | |
GZMM | |
Affinity chromatography | |
RUO | |
3004 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
4B2G4 | |
PE | |
GZMM | |
granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Granzyme M Monoclonal Antibody (4B2G4), PE, eBioscience™, Invitrogen™ > 100 Tests; PE
Spot an opportunity for improvement?Share a Content Correction