missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GRB2 (Human) Recombinant Protein
Human GRB2 full-length ORF ( AAH00631, 1 a.a. - 217 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002885-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEAIAKYDFKATADDELSFKRGDILKVLNEECDQNWYKAELNGKDGFIPKNYIEMKPHPWFFGKIPRAKAEEMLSKQRHDGAFLIRESESAPGDFSLSVKFGNDVQHFKVLRDGAGKYFLWVVKFNSLNELVDYHRSTSVSRNQQIFLRDIEQVPQQPTYVQALFDFDPQEDGELGFRRGDFIHVMDNSDPNWWKGACHGQTGMFPRNYVTPVNRNV
Specifications
AAH00631 | |
growth factor receptor-bound protein 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRB2 | |
GST |
2885 | |
Wheat germ expression system | |
25 μg | |
ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |