missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GRB2 (Human) Recombinant Protein
Human GRB2 full-length ORF ( AAH00631, 1 a.a. - 217 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH00631 |
---|---|
Gene ID (Entrez) | 2885 |
Name | growth factor receptor-bound protein 2 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16105731
|
Abnova™
H00002885-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16115731
|
Abnova™
H00002885-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MEAIAKYDFKATADDELSFKRGDILKVLNEECDQNWYKAELNGKDGFIPKNYIEMKPHPWFFGKIPRAKAEEMLSKQRHDGAFLIRESESAPGDFSLSVKFGNDVQHFKVLRDGAGKYFLWVVKFNSLNELVDYHRSTSVSRNQQIFLRDIEQVPQQPTYVQALFDFDPQEDGELGFRRGDFIHVMDNSDPNWWKGACHGQTGMFPRNYVTPVNRNV
Specifications
AAH00631 | |
growth factor receptor-bound protein 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2885 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRB2 | |
GST |