missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 Native Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

Brand:  Novus Biologicals™ DL-1207-NB

Product Code. 18072026

  • 3080.00 SEK / 0.50mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • A biologically active protein corresponding to Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4
  • Source: Griffonia (Bandeiraea) simplicifolia seeds
  • GSL I-B4 isolectin contains only the B subunits.
  • It is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit, and goat as well as a marker for non-peptidergic unmyelinated primary afferent neurons.
  • This “B”-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues.
  • DyLight™ 594 GSL I-B4 isolectin is produced by using special conjugation procedures to incorporate DyLight™ 594 into our affinity-purified lectin.
  • This conjugate has an appropriate number of fluorochromes bound which provide the maximum fluorescence and optimum staining characteristics for this particular lectin.
  • This reagent is supplied essentially free of unconjugated fluorochromes and inactive lectin.
  • Excitation maximum is at 592 nm and emission maximum is at 617 nm (red).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bioactivity, Immunocytochemistry/Immunofluorescence
Affinity purified
Store at 4°C in the dark.
10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM Ca2+
114 kDa
0.5 mg
DyLight 594
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 Native Protein > 0.5 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.