missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRO alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12335-1-AP-150UL
This item is not returnable.
View return policy
Description
The GRO gene was originally identified by subtractive hybridization studies between normal and tumorigenic Chinese hamster embryo fibroblasts. The hamster cDNA was cloned and used as a probe for cloning of the human GRO cDNA. The GROalpha gene initially cloned from T24 cells and the gene in melanoma cells encoding melanoma growth stimulating protein (MGSA) are identical.Specifications
GRO alpha | |
Polyclonal | |
Unconjugated | |
CXCL1 | |
C X C motif chemokine 1, CXCL1, FSP, GRO, GRO alpha(1 73), GRO1, GROa, Growth regulated alpha protein, MGSA, MGSA a, NAP 3, SCYB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2919 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09341 | |
CXCL1 | |
GRO alpha Fusion Protein Ag2917 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GRO alpha Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction