Learn More
GSDMB Mouse anti-Human, Unconjugated, Clone: 2F12C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67527-1-IG-20UL
This item is not returnable.
View return policy
Description
The GSDMB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSDMB. This antibody recognizes human antigen. The GSDMB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
GSDMB | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
2F12C4 | |
Unconjugated | |
20 μL | |
55876 | |
IgG1 |
Product Content Correction
Product Title
GSDMB Mouse anti-Human, Unconjugated, Clone: 2F12C4, Proteintech > 20 μL
Spot an opportunity for improvement?Share a Content Correction