Learn More
GSDMB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12885-1-AP-150UL
 View more versions of this product
Product Code. 16840834
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The GSDMB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GSDMB. This antibody recognizes human antigen. The GSDMB antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
GSDMB
Polyclonal
Unconjugated
150 μL
55876
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL
Rabbit
Primary
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GSDMB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL