Learn More
GSK3B Mouse anti-Human, Unconjugated, Clone: 2E9E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67329-1-IG-20UL
 View more versions of this product
Product Code. 16889895
  • 1645.54 SEK / 20µL
Description
The GSK3B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSK3B. This antibody recognizes human, pig antigen. The GSK3B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
GSK3B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA
2E9E4
Unconjugated
20 μL
2932
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GSK3B Mouse anti-Human, Unconjugated, Clone: 2E9E4, Proteintech > 20 μL