Learn More
GSTT1 Mouse anti-Human, Unconjugated, Clone: 1A4C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67645-1-IG-20UL
Description
The GSTT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTT1. This antibody recognizes human antigen. The GSTT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
GSTT1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1A4C11 | |
Unconjugated | |
20 μL | |
2952 | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed.