Learn More
GTF2A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10540-1-AP-150UL
 View more versions of this product
Product Code. 16886983
  • 4652.90 SEK / 150µL
Description
Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A), TFIID TFIIE TFIIF TFIIG/TFIIJ, and TFIIH respectively, and a 12-kD gamma subunit encoded by GTF2A2.
Specifications
Specifications
GTF2A2
Polyclonal
Unconjugated
GTF2A2
GTF2A2, HsT18745, TF2A2, TFIIA, TFIIA 12, TFIIA gamma, TFIIA p12 subunit, TFIIAS
Rabbit
Antigen Affinity Chromatography
RUO
235459, 2958, 83828
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08950, P52657, Q80ZM7
Gtf2a2
GTF2A2 Fusion Protein Ag0825
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GTF2A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated