missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2F2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15139-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.Specifications
GTF2F2 | |
Polyclonal | |
Unconjugated | |
GTF2F2 | |
ATP dependent helicase GTF2F2, BTF4, GTF2F2, RAP30, TF2F2, TFIIF, TFIIF beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2963, 68705, 81674 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13984, Q01750, Q8R0A0 | |
Gtf2f2 | |
GTF2F2 Fusion Protein Ag7313 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |