missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUCY1A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12605-1-AP-20UL
This item is not returnable.
View return policy
Description
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase, a soluble form (GCS or sGC), which act as receptors for nitric oxide and a membrane-bound receptor form (GC), which are peptide hormone receptors. The GC-C protein is composed of an extracellular domain, a single transmembrane domain, and a cytoplasmic region consisting of a kinase-like domain and a catalytic domain. It is expressed as two differentially glycosylated forms, a 130 kDa precursor form present in the endoplasmic reticulum and a 145 kDa form present on the plasma membrane. Ligand binding to the extracellular domain of GC-C promotes the accumulation of cGMP. GC-C acts as the receptor for heatstable enterotoxins, small peptides secreted by some pathogenic strains of E. coli that cause severe secretory diarrhea. GC-C also binds to guanylin and uroguanylin peptides, which modulate renal function in response to oral salt load.Specifications
GUCY1A3 | |
Polyclonal | |
Unconjugated | |
GUCY1A3 | |
GC SA3, GCS alpha 1, GCS alpha 3, GUC1A3, GUCA3, GUCSA3, GUCY1A1, GUCY1A3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2982, 497757, 60596 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19686, Q02108, Q9ERL9 | |
Gucy1a3 | |
GUCY1A3 Fusion Protein Ag3293 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GUCY1A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction