Learn More
HADHB Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29091-1-AP-150UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17221426
  • 4721.67 SEK / 150µL
Estimated Shipment: 28-05-2024
to see stock.
Description
Description
The HADHB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HADHB. This antibody recognizes human, mouse, rat antigen. The HADHB antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
HADHB
Polyclonal
Unconjugated
150 μL
3032
Liquid
Western Blot, ELISA
350 μg/mL
Rabbit
Primary
Human, Mouse, Rat