missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HADHB Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HADHB |
---|---|
Concentration | 350 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17231426
|
Proteintech
29091-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17221426
|
Proteintech
29091-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HADHB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HADHB. This antibody recognizes human, mouse, rat antigen. The HADHB antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
HADHB | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
3032 |
350 ÎĽg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse, Rat | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HADHB Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech