Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
Brand:  Hamilton™ 59000-10
Additional Details : Weight : 0.02500kg
 View more versions of this product
Product Code. 15132644
  • 1720.00 SEK / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Specifications
1702 BFP
25 μL
Microlab™ 600 dispensers