Learn More
RETSCH Hardened Stainless Steel and Tungsten Carbide O-ring
2675.00 SEK - 4615.00 SEK
Specifications
Product Type | O-Ring |
---|---|
Material | Hardened Stainless Steel and Tungsten Carbide |
Quantity | 1Each |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10111842
|
RETSCH
05.114.0056 |
For Planetary Ball Mill PM 100 CM 125mL Grinding Jars |
2675.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
11765635
|
RETSCH
05.114.0057 |
For Planetary Ball Mill PM 100 CM 50mL Grinding Jars |
4615.00 SEK
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
11722523
|
RETSCH
05.114.0055 |
For Planetary Ball Mill PM 100 CM 250mL Grinding Jars |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
O-Ring | |
Hardened Stainless Steel and Tungsten Carbide | |
1Each |