missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hCG beta Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11615-1-AP-150UL
This item is not returnable.
View return policy
Description
Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregcy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones includes pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. Alpha subunit, which is common to all the members of the family within a given species, with distinct but homologous beta subunits which confer hormone specificities.Specifications
hCG beta | |
Polyclonal | |
Unconjugated | |
CGB3 | |
CG beta, CGB, CGB3, CGB5, CGB7, CGB8, hCG, hCGB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1082 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0DN86 | |
CGB3 | |
hCG beta Fusion Protein Ag2191 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
hCG beta Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction