missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCLS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25003-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
HCLS1 | |
Polyclonal | |
Unconjugated | |
HCLS1 | |
CTTNL, HCLS1, HS1, LckBP1, p75 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3059 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14317 | |
HCLS1 | |
HCLS1 Fusion Protein Ag8645 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HCLS1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction