missing translation for 'onlineSavingsMsg'
Learn More

HDAC7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26207-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884725

  • 1355.00 SEK / 20µL
Estimated Shipment: 28-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Protein kinase D2 (PKD2) is a novel human serine threonine protein kinase gene. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKCmu and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.
Specifications

Specifications

HDAC7
Polyclonal
Unconjugated
HDAC7
HD7, HD7A, HDAC7, HDAC7A, histone deacetylase 7, Histone deacetylase 7A
Rabbit
Antigen Affinity Chromatography
RUO
51564
-20°C
Liquid
Immunoprecipitation
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WUI4
HDAC7
HDAC7 Fusion Protein Ag24396
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers