missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26207-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Protein kinase D2 (PKD2) is a novel human serine threonine protein kinase gene. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKCmu and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.Specifications
HDAC7 | |
Polyclonal | |
Unconjugated | |
HDAC7 | |
HD7, HD7A, HDAC7, HDAC7A, histone deacetylase 7, Histone deacetylase 7A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51564 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WUI4 | |
HDAC7 | |
HDAC7 Fusion Protein Ag24396 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |