Learn More
HDLBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15406-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
HDLBP | |
Polyclonal | |
Unconjugated | |
HDLBP | |
HBP, HDL binding protein, HDLBP, PRO2900, VGL, Vigilin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110611, 3069, 64474 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q00341, Q8VDJ3, Q9Z1A6 | |
Hdlbp | |
HDLBP Fusion Protein Ag7666 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
HDLBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction