Learn More
Gestigkeit Harry Heating Unit
Each SG hotplate is equipped with an output controller allowing variable adjustment of the heating output from 10 to 100%.
Brand:  Gestigkeit Harry SG4
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10330992
  • 16135.00 SEK / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
  • Series and supporting rod hot plates
  • Smoothly finished, cast-iron, hot plate with concave central expansion recess for warp free operation. All housing parts are constructed from stainless steel and each unit has a screw-on fitting for a supporting rod.
  • The appliances have been specifically designed for non-stop use.
  • Regulation of model
    • Each unit has a heating output controller that enables a variable adjustment between 10 to 100%.
Specifications
Specifications
4
705 x 210 x 160 mm
Stainless Steel
500°C
2000 W
Hotplate
230 V
Cast Steel
10.0 kg
50/60 Hz