missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HELT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24730-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGCG-3′.Specifications
HELT | |
Polyclonal | |
Unconjugated | |
HELT | |
HCM1228, HELT, HESL, Mgn | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234219, 391723, 498633 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A6NFD8, Q7TS99 | |
HELT | |
HELT Fusion Protein Ag20618 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HELT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction