Learn More
HESX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17927-1-AP-150UL
 View more versions of this product
Product Code. 16887934
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a conserved homeobox protein that is a transcriptional repressor in the developing forebrain and pituitary gland. Mutations in this gene are associated with septooptic dysplasia, HESX1-related growth hormone deficiency, and combined pituitary hormone deficiency.
Specifications
Specifications
HESX1
Polyclonal
Unconjugated
HESX1
ANF, hAnf, HESX homeobox 1, HESX1, Homeobox protein ANF, RPX
Rabbit
Antigen Affinity Chromatography
RUO
15209, 498575, 8820
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q61658, Q9UBX0
Hesx1
HESX1 Fusion Protein Ag12348
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HESX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated