missing translation for 'onlineSavingsMsg'
Learn More

HIF1AN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10646-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16897153

  • 1656.73 SEK / 20µL
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Factor Inhibiting HIF1 (FIH) protein facilitates repression of HIF-1 transcriptional activity by binding to VHL, which acts as a transcriptional corepressor. VHL inhibits HIF-1 alpha transactivation function by recruiting histone deacetylases. Involvement of VHL in association with FIH provides a unifying mechanism for the modulation of HIF-1 alpha protein stabilization and transcriptional activation in response to changes in cellular oxygen concentration.
Specifications

Specifications

HIF1AN
Polyclonal
Unconjugated
HIF1AN
Factor inhibiting HIF 1, FIH 1, FIH1, HIF1AN
Rabbit
Antigen Affinity Chromatography
RUO
309434, 319594, 55662
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BLR9, Q9NWT6
Hif1an
HIF1AN Fusion Protein Ag0997
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers