missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1AN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HIF1AN |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897153
|
Proteintech
10646-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16887153
|
Proteintech
10646-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Factor Inhibiting HIF1 (FIH) protein facilitates repression of HIF-1 transcriptional activity by binding to VHL, which acts as a transcriptional corepressor. VHL inhibits HIF-1 alpha transactivation function by recruiting histone deacetylases. Involvement of VHL in association with FIH provides a unifying mechanism for the modulation of HIF-1 alpha protein stabilization and transcriptional activation in response to changes in cellular oxygen concentration.Specifications
HIF1AN | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q8BLR9, Q9NWT6 | |
309434, 319594, 55662 | |
HIF1AN Fusion Protein Ag0997 | |
Primary | |
-20°C | |
HIF1AN |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Factor inhibiting HIF 1, FIH 1, FIH1, HIF1AN | |
Hif1an | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |