missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HISPPD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HISPPD1 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884545
|
Proteintech
26018-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874545
|
Proteintech
26018-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inositol phosphates.Specifications
HISPPD1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O43314 | |
23262 | |
HISPPD1 Fusion Protein Ag23374 | |
Primary | |
-20°C | |
PPIP5K2 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HISPPD1, hsVIP2, InsP6 and PP IP5 kinase 2, IP7K2, KIAA0433, PPIP5K2, VIP1 homolog 2, VIP2 | |
PPIP5K2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HISPPD1 Rabbit anti-Human, Polyclonal, Proteintech