missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST2H2AA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15302-1-AP-150UL
This item is not returnable.
View return policy
Specifications
HIST2H2AA4 | |
Polyclonal | |
Unconjugated | |
HIST2H2AA4 | |
HIST2H2AA4 | |
HIST2H2AA4 Fusion Protein Ag7560 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
H2A/R, H2AFO, HIST2H2AA, HIST2H2AA3, HIST2H2AA4, histone cluster 2, H2aa4, Histone H2A type 2 A, Histone H2A.2, Histone H2A/o | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
723790 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HIST2H2AA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction