missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Histone H2B |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895234
|
Proteintech
15857-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885234
|
Proteintech
15857-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transcription regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.Spezifikation
Histone H2B | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q16778, Q64524 | |
295274, 319190, 8349 | |
Histone H2B Fusion Protein Ag7811 | |
Primary | |
-20°C | |
HIST2H2BE |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GL105, H2B, H2B.1, H2B/q, H2BFQ, HIST2H2BE, histone cluster 2, H2be, Histone H2B, Histone H2B GL105, Histone H2B type 2 E, Histone H2B.q | |
Hist2h2be | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
Histone H2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech