Learn More
Cytiva HiTrap™ Capto™ AVB
Product Code. 16227791 Shop All Cytiva Products
Click to view available options
Capacity:
1 mL
5 mL
Unit Size:
Each
Pack of 5
Product Code. 16227791
Brand: Cytiva 17372211
Capto™ AVB is based on a rigid base matrix, delivering excellent pressure-flow properties to AAV production
  • Efficient purification of AAV of several subclasses by affinity chromatography
  • Prepacked HiTrap™ columns for easy screening and convenient process development, as well as small scale purification
  • High selectivity and excellent scalability
  • The possibility to run at higher flow rates and bed heights increases flexibility in process design and increases productivity
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.7 cm
Quantity 5/Pk.
Height (Metric) 2.5 cm
Flow Rate <4 mL/min (Max)
Content And Storage Store at 4°C to 8°C
Capacity 1 mL
Column Type Affinity Column
Shipping Condition Ambient
For Use With (Application) Easy Screening, Convenient Process Development, and Small Scale Purifications
Matrix Capto AVB
compliance-icons
Product Identifier
  • HiTrap 5x1ml Capto AVB
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.