missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21246-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
HMGB3L1 | |
Polyclonal | |
Unconjugated | |
HMGB3P1 | |
HMGB3P1 | |
HMGB3L1 Fusion Protein Ag15748 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HMG4L, HMGB3L1, HMGB3P1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
128872 | |
-20°C | |
Liquid |