missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HMGB3L1 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850865
|
Proteintech
21246-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840865
|
Proteintech
21246-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HMGB3L1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
HMG4L, HMGB3L1, HMGB3P1 | |
HMGB3P1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
128872 | |
HMGB3L1 Fusion Protein Ag15748 | |
Primary | |
-20°C | |
HMGB3P1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HMGB3L1 Rabbit anti-Human, Polyclonal, Proteintech