missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HNRNPD |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890664
|
Proteintech
12770-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880664
|
Proteintech
12770-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hnRNP D/AUF1 binds with high affinity to RNA molecules that contain AU-rich elements (AREs) found within the 3′ untranslated regions of many protooncogenes and cytokine mRNAs. It also binds to double and single stranded DNA sequences in a specific manner and functions a transcription factor. Each of the RNA-binding domains specifically can bind solely to a single stranded nonmonotonous 5′-UUAG-3′ sequence and also weaker to the single-stranded 5′-TTAGGG-3′ telomeric DNA repeat. It may be involved in translationally coupled mRNA turnover (referenced from swissprot).Specifications
HNRNPD | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q14103, Q60668, Q9JJ54 | |
11991, 3184, 79256 | |
HNRNPD Fusion Protein Ag3458 | |
Primary | |
-20°C | |
HNRNPD |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AUF1, AUF1A, hnRNP D0, HNRNPD, hnRNPD0, HNRPD, P37 | |
HNRNPD | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HNRNPD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech