Learn More
HNRNPH1 Mouse anti-Human, Unconjugated, Clone: 1A6C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67375-1-IG-150UL
 View more versions of this product
Product Code. 16879965
  • 4652.90 SEK / 150µL
Description
The HNRNPH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HNRNPH1. This antibody recognizes human, mouse, rat antigen. The HNRNPH1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
HNRNPH1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1A6C2
Unconjugated
150 μL
3187
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HNRNPH1 Mouse anti-Human, Unconjugated, Clone: 1A6C2, Proteintech > 150 μL