Learn More
HNRNPUL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10578-1-AP-150UL
 View more versions of this product
Product Code. 16887043
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This antibody can recognize isoform 1-4.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55 kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55 kDa in adenovirus-infected cells. This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55 kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55 kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.
Specifications
Specifications
HNRNPUL1
Polyclonal
Unconjugated
HNRNPUL1
E1B AP5, E1BAP5, HNRNPUL1, HNRPUL1
Rabbit
Antigen Affinity Chromatography
RUO
11100, 232989, 361522
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VDM6, Q9BUJ2
Hnrnpul1
HNRNPUL1 Fusion Protein Ag0899
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HNRNPUL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated