Learn More
HNRNPUL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10578-1-AP-150UL
 View more versions of this product
Product Code. 16887043
  • 4652.90 SEK / 150µL
Description
This antibody can recognize isoform 1-4.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55 kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55 kDa in adenovirus-infected cells. This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55 kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55 kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.
Specifications
Specifications
HNRNPUL1
Polyclonal
Unconjugated
HNRNPUL1
E1B AP5, E1BAP5, HNRNPUL1, HNRPUL1
Rabbit
Antigen Affinity Chromatography
RUO
11100, 232989, 361522
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VDM6, Q9BUJ2
Hnrnpul1
HNRNPUL1 Fusion Protein Ag0899
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HNRNPUL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated