missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hoechst 33258 analog 3

MedChemExpress Hoechst 33258 analog 3 are part of a family of blue fluorescent dyes used to stain DNA.

Encompass_Preferred

5632.94 SEK - 36162.84 SEK

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Solubility DMSO : 3.57 mg/mL (8.45 mM; ultrasonic and warming and heat to 70°C)
Molecular Weight (g/mol) 422.52 g/mol
CAS 23554-98-5
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16496846
Encompass_Preferred
View Documents
MedChemExpress
HY-15625
100 mg
36162.84 SEK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16482777
Encompass_Preferred
View Documents
MedChemExpress
HY-15625
50 mg
24118.94 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16466698
Encompass_Preferred
View Documents
MedChemExpress
HY-15625
10 mg
9009.59 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16439257
Encompass_Preferred
View Documents
MedChemExpress
HY-15625
5 mg
5632.94 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
DMSO : 3.57 mg/mL (8.45 mM; ultrasonic and warming and heat to 70°C)
23554-98-5
Room temperature
422.52 g/mol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MedChemExpress Hoechst 33258 analog 3

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.