Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
655.17 SEK - 1008.93 SEK
Product Code | Brand | Length (Metric) | Width (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
10331575
|
Hoefer™
TE46 |
14.5 cm | 21.5 cm | TE42 or TE62 Tank Blotting Units |
1008.93 SEK
Pack of 50 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
10762097
|
Hoefer™
TE76 |
21 cm | 25 cm | TE77 Semi-Dry Blotting Units |
771.65 SEK
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
11544434
|
Hoefer™
TE26 |
9 cm | 10.5 cm | TE22 Mini Tank Blotting Unit or MiniVE Blot Module |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12069847
|
Hoefer™
TE76 |
21 cm | 25 cm | TE77 Semi-Dry Blotting Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12014757
|
Hoefer™
TE46 |
14.5 cm | 21.5 cm | TE42 or TE62 Tank Blotting Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11544454
|
Hoefer™
TE76-1416 |
14 cm | 16 cm | TE70 or TE77 Semi-Dry Blotting Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10136664
|
Hoefer™
TE76-1416 |
14 cm | 16 cm | TE70 or TE77 Semi-Dry Blotting Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- For Western Blotting
Specifications
Sheet | |
Cellulose |