Learn More
Jaytec Glass™ Hoffman Voltameter Glass Unit
The Hoffman Voltameter Glass Unit is for classroom demonstration of electrolysis to determine the chemical composition of water.
Brand:  Jaytec Glass™ EKY-700-S
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12381429
  • 1520.00 SEK / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • Manufactured in borosilicate glass
  • Available Graduated or Ungraduated
  • H-pattern style
  • 50 mL capacity
Specifications
Specifications
Ungraduated Hoffman Voltameter
H-Pattern
To demonstrate electrolysis of water into oxygen and hydrogen
0 to 50 mL x 0.2 mL