missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11143-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein is a postsynaptic density scaffolding protein. Alternative splicing results in multiple transcript variants. Two related pseudogenes have been identified on chromosome 14.Specifications
HOMER2 | |
Polyclonal | |
Unconjugated | |
HOMER2 | |
ACPD, CPD, Cupidin, HOMER 2, homer homolog 2 (Drosophila), Homer protein homolog 2, HOMER2, HOMER2A, HOMER2B, Vesl 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9455 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NSB8 | |
HOMER2 | |
HOMER2 Fusion Protein Ag1606 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |