missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HOP2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888343
|
Proteintech
11339-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878343
|
Proteintech
11339-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMC3IP plays an important role in meiotic recombination. It stimulates DMC1-mediated strand exchange required for pairing homologous chromosomes during meiosis. The complex PSMC3IP/MND1 binds DNA, stimulates the recombinase activity of DMC1 as well as DMC1 D-loop formation from double-strand DNA. This complex stabilizes presynaptic RAD51 and DMC1 filaments formed on single strand DNA to capture double-strand DNA. This complex stimulates both synaptic and presynaptic critical steps in RAD51 and DMC1-promoted homologous pairing. It may inhibit HIV-1 viral protein TAT activity and modulate the activity of proteasomes through association with PSMC3.Specifications
HOP2 | |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9P2W1 | |
29893 | |
HOP2 Fusion Protein Ag1883 | |
Primary | |
-20°C | |
PSMC3IP |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GT198, HOP2, HUMGT198A, PSMC3 interacting protein, PSMC3IP, TBP 1 interacting protein, TBPIP | |
PSMC3IP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HOP2 Rabbit anti-Human, Polyclonal, Proteintech