Läs mer
Medicom™ Hopen™ Simple White Coveralls
Medicom™ Hopen™ Simple White Coveralls available in a variety of sizes, and styles to fit every need
Brand: Medicom™ 40.101/04
Beskrivning
- Single use, white coveralls can be used for a wide variety of industrial needs
- Individually packaged for hygiene, and controlled distribution purposes
- Suitable to HACCP procedures
Specifikationer
| X-Large | |
| Polypropylene | |
| Yes | |
| Elastic | |
| Collared | |
| Stitched | |
| Manufactured under an ISO 9001 Certified Quality System | |
| 40 g/m2 |
| Unisex | |
| White | |
| Elastic | |
| Elastic | |
| Zipper | |
| Non-sterile | |
| None |